Learn More
Buehler Release Agent, 4oz
Catalog No. NC1626234
Encompass
This item has a minimum qty of 10 per supplier requirements.
Liquid release agent for easier removal of mounts from castable molds or compression mounting presses.
Catalog No. NC1626234 Supplier Buehler Supplier No. 208186004
This item has a minimum qty of 10 per supplier requirements.
Liquid release agent for easier removal of mounts from castable molds or compression mounting presses.