Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Releasing Agent, Lanthanum Nitrate, 5% La in 5% HNO3, Ricca Chemical
SDP

Catalog No. MMLA50KN50 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
50 mL
Packaging:
Natural Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
MMLA50KN-50 50 mL Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MMLA50KN-50 Supplier Ricca Chemical Company Supplier No. MMLA50KN50
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Lanthanum nitrate releasing agent in 5% nitric acid is designed for use as a general laboratory reagent.

Specifications

Boiling Point 100°C
Chemical Name or Material Releasing Agent
CAS 7732-18-5, 1312-81-8, 7697-37-2
Color Colorless
Physical Form Liquid
Quantity 50 mL
UN Number UN3264
Solubility Information Miscible
Percent Purity 5%
Grade Laboratory
Concentration or Composition (by Analyte or Components) 5% La in 5% HNO3
Manufacturing Origin USA
Odor Odorless
Packaging Natural Poly Bottle
Appearance Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.