Learn More
Medchemexpress LLC Relebactam | 1174018-99-5 | 348.38 | 5 MG
SDP
Catalog No. 5000391238
Encompass_Preferred
Relebactam is a diazabicyclooctane inhibitor with activity against a wide spectrum of β-lactamases, including class A (extended-spectrum β-lactamases and KPC) and class C (AmpC) enzymes. It also demonstrates antibacterial activity.
  • Diazabicyclooctane inhibitor
  • Active against wide spectrum of β-lactamases
  • Shows antibacterial activity
  • Used in infection research
Catalog No. 50-003-91238 Supplier Medchemexpress LLC Supplier No. HY167525MG
Relebactam is a diazabicyclooctane inhibitor with activity against a wide spectrum of β-lactamases, including class A (extended-spectrum β-lactamases and KPC) and class C (AmpC) enzymes. It also demonstrates antibacterial activity.
  • Diazabicyclooctane inhibitor
  • Active against wide spectrum of β-lactamases
  • Shows antibacterial activity
  • Used in infection research