Learn More
Medchemexpress LLC Relenopride hydrochloride | 1221416-42-7 | 99.9% | 529.43 | 1 ML
SDP
Catalog No. 5000393677
Encompass_Preferred
Relenopride hydrochloride is a specific and selective 5-HT4 receptor agonist, with a Ki of 4.96 nM. It exhibits significantly lower affinity for 5-HT2A and 5-HT2B receptors compared to 5-HT4. This compound is known to increase gastrointestinal (GI) motility.
  • Specific and selective 5-HT4 receptor agonist
  • Increases gastrointestinal (GI) motility
  • Exhibits lower affinity for 5-HT2A and 5-HT2B receptors
Catalog No. 50-003-93677 Supplier Medchemexpress LLC Supplier No. HY16729A10MM/1ML
Relenopride hydrochloride is a specific and selective 5-HT4 receptor agonist, with a Ki of 4.96 nM. It exhibits significantly lower affinity for 5-HT2A and 5-HT2B receptors compared to 5-HT4. This compound is known to increase gastrointestinal (GI) motility.
  • Specific and selective 5-HT4 receptor agonist
  • Increases gastrointestinal (GI) motility
  • Exhibits lower affinity for 5-HT2A and 5-HT2B receptors