Learn More
Anton Paar Reliable measurement of standard solid samplesAccurate results for samples from 4.5 cm3 to 135 cm3Flow or pulse sample preparation modes
Catalog No. 5000580534
Encompass_Preferred
Performance specificationsAccuracy: 0.02 %Number reference volumes: 2 (TruPyc)Nominal reference volumes: 50 cm3 & 8 cm3Inserts available: 135 cm3, 50 cm3, 10 cm3, 4.5 cm3, 1.8 cm3, 0.25 cm3Preparation modes: Flow, pulse, noneGas expansion direction: Sample chamber firstSample chamber closure: Repeatable dual-turn self-aligning lid (TruLock)Display & control:7-inch TFT WVGA (800×480) PCAP touchscreenGraphical user interfaceTransducer accuracy: Better than 0.1 %Pressure reading resolution: 0.0001 psiDigital pressure readout: 0.00001 bar
Catalog No. 50-005-80534 Supplier Anton Paar Supplier No. 231522
Please contact your sales representative for a quote or further ordering instructions.
Performance specificationsAccuracy: 0.02 %Number reference volumes: 2 (TruPyc)Nominal reference volumes: 50 cm3 & 8 cm3Inserts available: 135 cm3, 50 cm3, 10 cm3, 4.5 cm3, 1.8 cm3, 0.25 cm3Preparation modes: Flow, pulse, noneGas expansion direction: Sample chamber firstSample chamber closure: Repeatable dual-turn self-aligning lid (TruLock)Display & control:7-inch TFT WVGA (800×480) PCAP touchscreenGraphical user interfaceTransducer accuracy: Better than 0.1 %Pressure reading resolution: 0.0001 psiDigital pressure readout: 0.00001 bar