Learn More
Grainger Relief Boot RJ45 White PK50
Catalog No. 502749844
Encompass_Preferred
The RELIEF BOOT RJ45 WHITE PK-50 is a pack of 50 white RJ45 boots from RELIEF. These boots are designed to protect unused RJ45 ports from dust and debris. They are made from durable ABS plastic and have flexible tabs that allow for easy installation and removal. The boots have a universal fit and are compatible with CAT5, CAT5e, CAT6, and CAT6a network cables. They meet EIA/TIA 568B standards. This pack contains 50 individual boots so provides enough to protect multiple unused ports on networking equipment, patch panels, and wall plates. The boots help maintain signal integrity and prevent network issues that can occur from exposed ports collecting dust and dirt.
Catalog No. 50-274-9844 Supplier Grainger Supplier No. 14J349
The RELIEF BOOT RJ45 WHITE PK-50 is a pack of 50 white RJ45 boots from RELIEF. These boots are designed to protect unused RJ45 ports from dust and debris. They are made from durable ABS plastic and have flexible tabs that allow for easy installation and removal. The boots have a universal fit and are compatible with CAT5, CAT5e, CAT6, and CAT6a network cables. They meet EIA/TIA 568B standards. This pack contains 50 individual boots so provides enough to protect multiple unused ports on networking equipment, patch panels, and wall plates. The boots help maintain signal integrity and prevent network issues that can occur from exposed ports collecting dust and dirt.