Learn More
AdipoGen anti-RELM-alpha (mouse), mAb (MREL 127) (50 µg)
SDP
Supplier:  AdipoGen AG20A0060C050
Encompass_Preferred
Monoclonal Antibody. Recognizes mouse RELM-alpha. Detects a band of ~10kDa by Western blot. Isotype: Rat IgG2akappa. Clone: MREL 127. Applications: ELISA, WB. Liquid. 0.2µm-filtered solution in PBS, pH 7.4. Contains no preservatives. RELM-alpha is a member to the family of adipocyte-secreted proteins called adipocytokines. It is abundantly expressed in white adipose tissue but not in preadipocytes, nor 3T3-L1 adipocytes. RELM-alpha is involved in the regulation of colonic inflammation and might be a link between colonic injury, glucose tolerance, and energy intake.
Catalog No. 50-205-2859