Learn More
AdipoGen RELM-beta (human) Matched Pair Detection Set (1 Set)
SDP
Catalog No. 502053331
Encompass_Preferred
Matched Pair Detection Set. Detects human RELM-beta. Does not cross-react with human adiponectin, mouse adiponectin, human RBP4, human Nampt, human vaspin, human GPX3, human ANGPTL3, human progranulin, human leptin, human resistin, human IL-33, human sirtuin 1, human clusterin, mouse resistin, human FABP4. Colorimetric assay. Sample Types: . Range: 0.016 to 1ng/ml. Sensitivity: 10pg/ml. RELM-beta has been suggested to play a regulatory role during inflammation and may also act to establish links among adipose tissue, the intestine and the liver. Interestingly, the molecular structure of RELM-beta is highly homologous to that of the adipose-derived cytokine resistin. The specific and sensitive RELM-beta Matched Pair Detection Set might detect RELM-beta in serum of gut-inflamed patients. Natural levels of RELM-beta were seen in stool, but were not detectable in the serum or other cell culture models of healthy human patients. Further testing are ongoing.
Catalog No. 50-205-3331 Supplier AdipoGen Supplier No. AG46A0001KI01
Matched Pair Detection Set. Detects human RELM-beta. Does not cross-react with human adiponectin, mouse adiponectin, human RBP4, human Nampt, human vaspin, human GPX3, human ANGPTL3, human progranulin, human leptin, human resistin, human IL-33, human sirtuin 1, human clusterin, mouse resistin, human FABP4. Colorimetric assay. Sample Types: . Range: 0.016 to 1ng/ml. Sensitivity: 10pg/ml. RELM-beta has been suggested to play a regulatory role during inflammation and may also act to establish links among adipose tissue, the intestine and the liver. Interestingly, the molecular structure of RELM-beta is highly homologous to that of the adipose-derived cytokine resistin. The specific and sensitive RELM-beta Matched Pair Detection Set might detect RELM-beta in serum of gut-inflamed patients. Natural levels of RELM-beta were seen in stool, but were not detectable in the serum or other cell culture models of healthy human patients. Further testing are ongoing.