Learn More
AdipoGen RELM-beta human pAb
SDP
Supplier:  AdipoGen AG25A0012C100
Encompass_Preferred
Polyclonal Antibody. Recognizes human RELM-beta. Detects a band of ~19kDa by Western blot. Does not detect human Resistin, mouse Resistin, mouse RELM-alpha, mouse RELM-beta or mouse RELM-gamma. Source Rabbit. Applications ELISA, IHC, WB. Liquid. 0.2µm-filtered solution in PBS, pH 7.4. Contains no preservatives. RELM-beta is a 19kDa disulfide-linked homodimeric protein expressed in the epithelium of the colon and small bowel. RELM-beta has been suggested to play a regulatory role during inflammation and may also act to establish links among adipose tissue, the intestine and the liver. Interestingly, the molecular structure of RELM-beta is highly homologous to that of the adipose-derived cytokine resistin.
Catalog No. 50-256-3300