Learn More
Medchemexpress LLC Relugolix-d6 | 737789-87-6 | 98.8% | 629.67 | 1 MG
SDP
Catalog No. 5000391834
Encompass_Preferred
Relugolix-d6 is deuterium labeled Relugolix, a potent, orally active, nonpeptidic gonadotropin-releasing hormone (GnRH) antagonist. It possesses high affinity and potent antagonistic activity for human and monkey receptors and is used for the study of sex-hormone-dependent diseases, including endometriosis, uterine fibroids, and prostate cancer.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-003-91834 Supplier Medchemexpress LLC Supplier No. HY16474S1MG
Relugolix-d6 is deuterium labeled Relugolix, a potent, orally active, nonpeptidic gonadotropin-releasing hormone (GnRH) antagonist. It possesses high affinity and potent antagonistic activity for human and monkey receptors and is used for the study of sex-hormone-dependent diseases, including endometriosis, uterine fibroids, and prostate cancer.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS