Learn More
Chemglass Life Sciences Condenser, Reflux, 24/40 Joint, 210 x 64mm, GL-14 Screw Caps, Removable Hose Connections
SDP
Catalog No. CG1217HC20
Encompass_Preferred
Condenser, Reflux, 24/40 Joint, 210 x 64mm, GL-14 Screw Caps, Removable Hose Connections. Compact high efficiency reflux condenser similar to CG-1217 but with detachable hose connections. Connections are made of polypropylene and provide a safe and easy way of connecting and disconnecting flexible tubing to the condenser. Supplied complete with two GL open top screw caps and two straight polypropylene hose connections. Condensing area is the same as CG-1217.
Catalog No. CG1217HC20 Supplier Chemglass Life Sciences Supplier No. CG1217HC20
Condenser, Reflux, 24/40 Joint, 210 x 64mm, GL-14 Screw Caps, Removable Hose Connections. Compact high efficiency reflux condenser similar to CG-1217 but with detachable hose connections. Connections are made of polypropylene and provide a safe and easy way of connecting and disconnecting flexible tubing to the condenser. Supplied complete with two GL open top screw caps and two straight polypropylene hose connections. Condensing area is the same as CG-1217.