Learn More
Chemglass Life Sciences Condenser, 29/42 Joint, Reflux, 230 x 65mm
SDP
Catalog No. CG121721
Encompass_Preferred
Condenser, 29/42 Joint, Reflux, 230 x 65mm. Compact high efficiency reflux condenser employs a double wall thimble shape bulb sealed into the outer jacket. Serrated hose connections for water connection extend through the outer bulb and have an O.D. of 10mm at the middle serration. While the overall height of this condenser is only 210-215mm, it has the efficiency of a standard 400mm Liebig or West condenser.
Catalog No. CG121721 Supplier Chemglass Life Sciences Supplier No. CG121721
Condenser, 29/42 Joint, Reflux, 230 x 65mm. Compact high efficiency reflux condenser employs a double wall thimble shape bulb sealed into the outer jacket. Serrated hose connections for water connection extend through the outer bulb and have an O.D. of 10mm at the middle serration. While the overall height of this condenser is only 210-215mm, it has the efficiency of a standard 400mm Liebig or West condenser.