Learn More
Aviva Systems Biology Corp REM1 Antibody - N-terminal region (ARP54963_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP54963_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against REM1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human REM1. Alternate names: GD:REM, GES, MGC48669, REMApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep, Yeast, Zebrafish
Catalog No. 50-014-53877