Learn More
Description
Remazol Brilliant Blue R is a hydrophobic, anthraquinone dye which has several commercial uses.
Specifications
Specifications
| Chemical Name or Material | Remazol Brilliant Blue R |
| CAS | 2580-78-1 |
| Color | Black to Blue |
| Quantity | 25 g |
| Molecular Weight (g/mol) | 626.54 g/mol |
| Percent Purity | 40 to 65% |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.