Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ REMBRANDT™ DISH & AP (DIG-AP) Detection Kit, for HPV typing

Catalog No. A103K9905
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A103K9905 1 kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A103K9905 Supplier Thermo Scientific™ Supplier No. A103K.9905
  • $1,596.00
    $1,197.65 / Each of 1

    Save $398.35

    25% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

REMBRANDT™ DISH & AP (DIG-AP) Detection Kit, for HPV typing

REMBRANDT™ DISH & AP (DIG-AP) Detection Kit, for HPV typing

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store in refrigerator (2°C to 8°C).
Detection Method Colorimetric
Label or Dye AP, Digoxigenin
Product Line REMBRANDT
Product Type DISH AP Detection Kit
Quantity 1 kit
Shipping Condition Wet Ice
Target Human Papillomavirus (HPV)
Substrate BCIP/NBT
Target Molecule DNA
Target Specificity Target-Specific
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.