Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ REMBRANDT™ RISH & HRP (DIG-HRP) Detection Kit, for Epstein-Barr encoded early RNAs (EBER)

Catalog No. A500K9901
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A500K9901 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A500K9901 Supplier Thermo Scientific™ Supplier No. A500K.9901
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

REMBRANDT™ RISH & HRP (DIG-HRP) Detection Kit, for Epstein-Barr encoded early RNAs (EBER)

REMBRANDT™ RISH & HRP (DIG-HRP) Detection Kit, for Epstein-Barr encoded early RNAs (EBER)

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store in refrigerator (2°C to 8°C).
Detection Method Colorimetric
Label or Dye Digoxigenin, HRP
Product Line REMBRANDT
Product Type RISH HRP Detection Kit
Quantity 1 Kit
Shipping Condition Wet Ice
Target Epstein-Barr Virus (EBV)
Substrate AEC
Target Molecule RNA
Target Specificity Target-Specific
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.