Learn More
Medchemexpress LLC HY-104077 1mg Medchemexpress, Remdesivir CAS:1809249-37-3 Purity:>98%
SDP
Catalog No. 501968206
Encompass_Preferred
Medchemexpress, HY-104077 1mg Remdesivir CAS:1809249-37-3 Remdesivir (GS-5734) is a nucleoside analogue, with effective antiviral activity, with EC50s of 74 nM for SARS-CoV and MERS-CoV in HAE cells, and 30 nM for murine hepatitis virus in delayed brain tumor cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8206 Supplier Medchemexpress LLC Supplier No. HY1040771MG
Medchemexpress, HY-104077 1mg Remdesivir CAS:1809249-37-3 Remdesivir (GS-5734) is a nucleoside analogue, with effective antiviral activity, with EC50s of 74 nM for SARS-CoV and MERS-CoV in HAE cells, and 30 nM for murine hepatitis virus in delayed brain tumor cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.