Learn More
TARGETMOL CHEMICALS INC REMDESIVIR 5MG
SDP
Catalog No. 502579126
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 200 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Remdesivir (GS-5734) is a nucleoside analogue with effective antiviral activity( EC50s of 74 nM for SARS-CoV and MERS-CoV in HAE cells)purity: 99%
Catalog No. 50-257-9126 Supplier TARGETMOL CHEMICALS INC Supplier No. T77665MG
Add to Cart
Add to Cart
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 200 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Remdesivir (GS-5734) is a nucleoside analogue with effective antiviral activity( EC50s of 74 nM for SARS-CoV and MERS-CoV in HAE cells)purity: 99%