Learn More
Cayman Chemical RemdesIvIr-d4 1mg
SDP
Supplier:  Cayman Chemical 309051 MG
Encompass_Preferred
An internal standard for the quantification of remdesivir by GC- or LC-MS
Catalog No. 50-299-0294