Learn More
Cayman Chemical RemdesIvIr-d4 500ug
SDP
Supplier:  Cayman Chemical 30905500 UG
Encompass_Preferred
An internal standard for the quantification of remdesivir by GC- or LC-MS
Catalog No. 50-299-1766