Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Remdesivir-d5 | 00-00-0 | MFCD31657351 | 99.9% | 607.61 g·mol⁻¹ | C27H30D5N6O8P | 5 MG
SDP

Catalog No. 5000244343
Encompass_Preferred

Remdesivir-d5 is a deuterium-labeled analogue of remdesivir supplied as a stable isotope standard for research and analytical applications, including quantitative mass spectrometry and antiviral studies.

  • Deuterium-labeled (d5) stable isotope standard for analytical assays.
  • High purity (99.9%) suitable for quantitative analysis.
  • Molecular weight ~607.61 g·mol⁻¹ and formula C27H30D5N6O8P.
  • Supplied as a 5 mg pack for laboratory use.
  • Intended for research use only; not for human therapeutic use.

Catalog No. 50-002-44343 Supplier Medchemexpress LLC Supplier No. HY104077S5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Remdesivir-d5 is a deuterium-labeled analogue of remdesivir supplied as a stable isotope standard for research and analytical applications, including quantitative mass spectrometry and antiviral studies.

  • Deuterium-labeled (d5) stable isotope standard for analytical assays.
  • High purity (99.9%) suitable for quantitative analysis.
  • Molecular weight ~607.61 g·mol⁻¹ and formula C27H30D5N6O8P.
  • Supplied as a 5 mg pack for laboratory use.
  • Intended for research use only; not for human therapeutic use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.