Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Remdesivir-d5 (GS-5734-d5) | 00-00-0 | 99.9% | 607.61 g·mol⁻1 | C27H30D5N6O8P | 1 MG
SDP

Catalog No. 5000242210
Encompass_Preferred

Remdesivir-d5 (GS-5734-d5) is a deuterium-labeled analogue of the antiviral nucleoside remdesivir, provided as an isotopic internal standard for analytical and research applications. It is intended for quantitative studies by LC-MS, GC-MS, and NMR and is supplied as a white to off-white solid.

  • Deuterium-labeled internal standard for quantitative analysis
  • High chemical purity and isotopic enrichment
  • Molecular formula C27H30D5N6O8P; molecular weight 607.61 g·mol⁻1
  • Supplied as a white to off-white solid with reported purity 99.9%
  • Storage: powder at -20°C for long-term, 4°C for short-term
  • For research use only; not for human use

Catalog No. 50-002-42210 Supplier Medchemexpress LLC Supplier No. HY104077S1MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Remdesivir-d5 (GS-5734-d5) is a deuterium-labeled analogue of the antiviral nucleoside remdesivir, provided as an isotopic internal standard for analytical and research applications. It is intended for quantitative studies by LC-MS, GC-MS, and NMR and is supplied as a white to off-white solid.

  • Deuterium-labeled internal standard for quantitative analysis
  • High chemical purity and isotopic enrichment
  • Molecular formula C27H30D5N6O8P; molecular weight 607.61 g·mol⁻1
  • Supplied as a white to off-white solid with reported purity 99.9%
  • Storage: powder at -20°C for long-term, 4°C for short-term
  • For research use only; not for human use

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.