Learn More
Apexbio Technology LLC Remdesivir (GS-5734), 5mg CAS# 1809249-37-3
SDP
Catalog No. 501998447
Encompass_Preferred
Remdesivir (GS-5734) is an antiviral nucleoside analogue that inhibits murine hepatitis virus (MHV) with similar EC50 as SARS-CoV and MERS-CoV. Coronaviruses (CoVs) are positive-sense, single-stranded RNA viruses that infect a wide range of animal hosts. In humans, CoVs were recognized as typically causing colds and pneumonia until the emergence of severe acute respiratory syndrome coronavirus (SARS-CoV) in 2002 and Middle East respiratory syndrome coronavirus (MERS-CoV) in 2012 from zoonotic sources. GS-5734 is the monophosphoramidate prodrug of the C-adenosine nucleoside analogue GS-441524. In delayed brain tumor (DBT) cells infected with MHV, GS-5734 inhibited MHV more potently than GS-441524, with a GS-5734 EC50 of 0.03 M. Furthermore, EC50 values were determined after treatment with GS-441524 and GS-5734 in SARS-CoV- and MERS-CoV-infected primary human airway epithelial cell (HAE) cultures. Other sizes are also available. Please inqury us for quote.
Catalog No. 50-199-8447 Supplier Apexbio Technology LLC Supplier No. B83985
Add to Cart
Add to Cart
Remdesivir (GS-5734) is an antiviral nucleoside analogue that inhibits murine hepatitis virus (MHV) with similar EC50 as SARS-CoV and MERS-CoV. Coronaviruses (CoVs) are positive-sense, single-stranded RNA viruses that infect a wide range of animal hosts. In humans, CoVs were recognized as typically causing colds and pneumonia until the emergence of severe acute respiratory syndrome coronavirus (SARS-CoV) in 2002 and Middle East respiratory syndrome coronavirus (MERS-CoV) in 2012 from zoonotic sources. GS-5734 is the monophosphoramidate prodrug of the C-adenosine nucleoside analogue GS-441524. In delayed brain tumor (DBT) cells infected with MHV, GS-5734 inhibited MHV more potently than GS-441524, with a GS-5734 EC50 of 0.03 M. Furthermore, EC50 values were determined after treatment with GS-441524 and GS-5734 in SARS-CoV- and MERS-CoV-infected primary human airway epithelial cell (HAE) cultures. Other sizes are also available. Please inqury us for quote.