Learn More
Medchemexpress LLC Remdesivir metabolite | 1911579-04-8 | 99.6% | C15H19N6O8P | 10MG
SDP
Catalog No. 5000260918
Encompass_Preferred
GS-704277 is an alanine metabolite of remdesivir used as a research reference standard for antiviral metabolism and analytical studies. It is intended for metabolite identification, pharmacokinetic investigations, and development or validation of chromatographic and mass-spectrometric assays.
  • High stated purity suitable for analytical applications.
  • Molecular formula and weight provided for confirmation and reporting.
  • Available in small research-scale pack sizes for bench studies.
  • Useful for metabolite identification and pharmacokinetic work.
  • Compatible with LC-MS and HPLC analytical workflows.
Catalog No. 50-002-60918 Supplier Medchemexpress LLC Supplier No. HY13630310MG
GS-704277 is an alanine metabolite of remdesivir used as a research reference standard for antiviral metabolism and analytical studies. It is intended for metabolite identification, pharmacokinetic investigations, and development or validation of chromatographic and mass-spectrometric assays.
  • High stated purity suitable for analytical applications.
  • Molecular formula and weight provided for confirmation and reporting.
  • Available in small research-scale pack sizes for bench studies.
  • Useful for metabolite identification and pharmacokinetic work.
  • Compatible with LC-MS and HPLC analytical workflows.