Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Remel™ 0.02% Polysorbate 80

Catalog No. R14503
Change view
Click to view available options
Quantity:
100 x 25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R14503 100 x 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. R14503 Supplier Thermo Scientific™ Supplier No. R14503
Add to Cart
Add to Cart
This item is not returnable. View return policy

Thermo Scientific™ Remel 0.02% Polysorbate 80 is for MIC dilution.

Thermo Scientific™ Remel 0.02% Polysorbate 80 is for MIC dilution.

TRUSTED_SUSTAINABILITY

Specifications

Description Polysorbate 80, 0.02%
Format G - 25mL
Product Type Buffer Solution
Quantity 100 x 25 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.