Description
Thermo Scientific™ Remel 10B Arginine Broth is used for transport and selective cultivation of Ureaplasma urealyticum and Mycoplasma hominis.
Specifications
Specifications
| Description | 10B Arginine Broth |
| Format | B - 1.8mL |
| Product Type | Microbiology Media |
| Quantity | 100/Cs. |