Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Lauryl Tryptose Broth (1X) w/DT

Catalog No. p-4524154
Change view
Click to view available options
Quantity:
20 x 10 mL
Format:
20mm x 150mm Durham Tube
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Format
R09447 20 x 10 mL 20mm x 150mm Durham Tube
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. R09447 Supplier Thermo Scientific™ Supplier No. R09447
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Detect coliforms in water, wastewater and foods using Lauryl Tryptose Broth (LST Broth).

Detect coliforms in water, wastewater, and foods using Thermo Scientific™ Remel™ Lauryl Tryptose Broth (LST Broth). Lauryl Tryptose Broth is recommended by the American Public Health Association (APHA), AOAC International (AOAC) and the U.S. Food and Drug Administration (USFDA) for the detection of coliforms in water, wastewater, and foods1-4. The production of gas in Lauryl Tryptose Broth serves as a presumptive test for the presence of coliforms in water and foods.

Mallman and Darby developed Lauryl Tryptose Broth5,6 and they evaluated a variety of wetting agents and determined that addition of sodium lauryl sulfate to selective media produced the best results for the detection of the coliform group.

  • Recommended by the APHA, AOAC and USFDA for the detection of coliforms in water, wastewater, and foods1-4
  • Selective medium - Selective agent sodium lauryl sulfate inhibits many organisms other than coliforms
  • Ready-to-use - Convenience of prepared media.
  • Easy to interpret - Turbidity in the broth and gas bubbles in the inverted Durham tube indicates positive fermentation reaction:
    • Positive test - Turbidity with gas production within 48 hours
    • Negative test - Turbidity with no gas production within 48 hours

This medium contains casein peptone that supplies nitrogen, carbon, and sulfur required for the bacterial growth. Sodium chloride provides essential electrolytes and maintains osmotic equilibrium. Lactose is the carbohydrate source fermented by coliforms. Dipotassium phosphate and monopotassium phosphate present in the medium serve as buffering agents.

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Industrial Reference: AOAC, BAM, COMPF, EPA, SMWW, USDA.
Description Lauryl Tryptose Broth, 1X
Format 20mm x 150mm Durham Tube
Product Type Microbiology Media
Quantity 20 x 10 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.