Description
Thermo Scientific™ Remel 1X LB Broth (Luria-Bertani) is used for cultivation and maintenance of Escherichia coli for genetic and molecular studies.
Specifications
Specifications
| Description | LB Broth, 1X |
| Format | BB - 500mL |
| Product Type | Microbiology Media |
| Quantity | 4 x 500 mL |