Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Skim Milk Medium, 2X

Catalog No. R064548 Shop All Thermo Scientific Products
Change view
Click to view available options
Format:
15mm x 103mm Tube
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Format
R064548 15mm x 103mm Tube
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. R064548 Supplier Thermo Scientific™ Supplier No. R064548
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Differentiate microorganisms based on coagulation and proteolysis of casein using 2X Skim Milk Medium.

Differentiate microorganisms based on coagulation and proteolysis of casein with Thermo Scientific™ Remel™ 2X Skim Milk Medium. Skim milk is used as a substrate in media to identify psychrotropic bacteria which are commonly associated with milk products1. Such bacteria produce proteolytic enzymes that often remain active after treatment with ultra-high temperature sterilization2. Skim milk is also included in clinical laboratories to cultivate and identify organisms such as Mycobacterium tuberculosis, Cornybacterium diphtheriae, Clostridium butyricum3.

  • Also used as additive in microbiological media
TRUSTED_SUSTAINABILITY

Specifications

Description Skim Milk Medium 2X
Format 15mm x 103mm Tube
Product Type Microbiology Media
Quantity 20 x 5 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.