Description
Thermo Scientific™ Remel 3 Day Arylsulfatase Broth, is a test for differentiation of Mycobacterium fortuitum complex from saprophytic mycobacteria.
Specifications
Specifications
| Description | Arylsulfatase Broth, 3 day test |
| Format | 15mm x 103mm Tube |
| Product Type | Microbiology Media |
| Quantity | 20 x 2 mL |