Learn More
Description
Thermo Scientific™ Remel A-7 Agar is used for isolation and differentiation of Ureaplasma and some species of Mycoplasma.
Specifications
Specifications
| Format | 15mm x 60mm Plate |
| Media Type | A-7 Agar |
| Product Type | Agar Plate |
| Quantity | 10/Pk. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.