Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Remel™ Agar (1.5%)

Catalog No. R08007 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
20/Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R08007 20/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. R08007 Supplier Thermo Scientific™ Supplier No. R08007
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Thermo Scientific™ Remel Agar (1.5%) is for agar overlay Kirby-Bauer.

Thermo Scientific™ Remel Agar (1.5%) is for agar overlay Kirby-Bauer.

TRUSTED_SUSTAINABILITY

Specifications

Description Agar 1.5%
Format 20mm x 113mm Tube
Product Type Agar
Quantity 20/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.