Learn More
Thermo Scientific™ Remel™ Agglutinating Sera, E. coli Polyvalent
Shop All Thermo Scientific ProductsDescription
Remel Agglutinating Sera, E. coli Polyvalent is used for serological identification of E. coli by slide and/or tube agglutination tests.
Specifications
Specifications
| Description | E. coli Polyvalent 4 |
| Detectable Analytes | E. coli |
| Type | Agglutinating Sera |
| Packaging Type | 2 mL/Vial |
| Quantity | 2 mL |
| Specificity | types O44, O112, O124, O142 |