Learn More
Thermo Scientific™ Remel™ Agglutinating Sera, Neisseria meningitidis
Shop All Thermo Scientific ProductsDescription
Remel Agglutinating Sera are used for serological identification of N. meningitidis by rapid slide agglutination and CIE procedures.
Specifications
Specifications
| Description | Group B |
| CE Marker | Yes |
| Detectable Analytes | N. meningitidis |
| Type | Neisseria Meningitidis Test Kit |
| Organism Group | Group B |
| Quantity | Each |
| Target Organism Class | Neisseria Meningitidis |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.