Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Remel™ Agglutinating Sera, Neisseria meningitidis

Catalog No. r30166901
Change view
Click to view available options
Description:
Group A
Group B
Group C
Group D
Group W135
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Description
R30166901 Group C
R30166801 Group A
R30167001 Group D
R30167501 Group B
R30167401 Group W135
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. R30166901 Supplier Thermo Scientific™ Supplier No. R30166901
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Used for serological identification of N. meningitidis by rapid slide agglutination and CIE procedures.

Remel Agglutinating Sera are used for serological identification of N. meningitidis by rapid slide agglutination and CIE procedures.

TRUSTED_SUSTAINABILITY

Specifications

Description Group C
CE Marker Yes
Detectable Analytes N. meningitidis
Type Neisseria Meningitidis Test Kit
Organism Group Group C
Quantity Each
Target Organism Class Neisseria Meningitidis
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.