Learn More
Thermo Scientific™ Remel™ Agglutinating Sera, Salmonella H
Catalog No. R30161601
Change view
Click to view available options
Description:
Polyvalent, Phase 2
Polyvalent, Phases 1 and 2
Rapid Diagnostic 3 (d, E, G, k)
Salmonella H (a)
Salmonella H (b)
Salmonella H (c )
Salmonella H (d)
Salmonella H (i)
Salmonella H (r )
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Description
R30161601 Salmonella H (i)
R30160401 Salmonella H (c )
R30160201 Salmonella H (a)
R30160301 Salmonella H (b)
R30160501 Salmonella H (d)
R30858501 Polyvalent, Phases 1 and 2
R30162201 Salmonella H (r )
R30160101 Rapid Diagnostic 3 (d, E, G, k)
R30163201 Polyvalent, Phase 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
This item is not returnable. View return policy
Catalog No. R30161601 Supplier Thermo Scientific™ Supplier No. R30161601
Add to Cart
Add to Cart
This item is not returnable. View return policy
Used for identification of flagellar H antigens of Salmonella strains by slide and/or tube agglutination test.
Remel Agglutinating Sera, Salmonella H is used for identification of flagellar H antigens of Salmonella strains by slide and/or tube agglutination test.
  • Select sera also for use as positive controls for stained Salmonella suspensions
TRUSTED_SUSTAINABILITY
Specifications
Control Sets Positive
Description Salmonella H (i)
Detectable Analytes Salmonella H Antigen
Type Agglutinating Sera
Packaging Type 2 mL/Vial
Quantity 2 mL
Target Organism Class Salmonella H