Learn More
Description
Serologically identify somatic O antigens of Salmonella strains by slide agglutination test using Agglutinating Sera, Salmonella O.
Serologically identify somatic O antigens of Salmonella strains by slide agglutination test for epidemiological and diagnostic purposes using Thermo Scientific™ Remel™ Agglutinating Sera, Salmonella O. The sera are absorbed to remove agglutinins for other Salmonella antigens and the paracolon α agglutinin. Salmonella Somatic Agglutinating Sera (ZC series) are intended for the identification of O antigens.
- Select sera also for use as positive controls for stained Salmonella suspensions
Specifications
Specifications
| Control Sets | Positive |
| Description | Factor 11 (group F) |
| Detectable Analytes | Salmonella O Antigen |
| Type | Agglutinating Sera |
| Organism Group | Group F |
| Packaging Type | 2 mL/Vial |
| Quantity | 2 mL |
| Target Organism Class | Salmonella O |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.