Learn More
Thermo Scientific™ Shigella dysenteriae Polyvalent (1-10) Agglutinating Sera
Shop All Thermo Scientific ProductsDescription
Identify Shigella cultures presumptively with slide agglutination technique using Shigella dysenteriae Polyvalent (1-10) Agglutinating Sera.
Presumptively identify Shigella cultures for epidemiological and diagnostic purposes using Thermo Scientific™ Remel™ Shigella dysenteriae Polyvalent (1-10) Agglutinating Sera. These sera are suitable for use in slide agglutination tests. Serological tests are based on the fact that antibodies produced in the serum in response to exposure to bacterial antigens will visibly agglutinate with bacteria carrying homologous antigens. Antisera provide serological identification only; full identification of an organism must be made in conjunction with biochemical testing.
Shigella dysenteriae can be differentiated on the basis of their reactions with specific antisera. Remel Shigella dysenteriae Polyvalent (1-10) Agglutinating Sera cover S. dysenteriae types 1 to 10. As Shigella can be confused with some non-motile serotypes of E. coli (Alkalescens‑Dispar group) an additional serum (R30164301) is available for differentiation.
- Ready to use: Supplied as Ready to use agglutinating serum.
- Easy to interpret: Visible agglutination with S. dysenteriae types 1 to 10. Control solution shows no visible agglutination.
Shigella dysenteriae Polyvalent (1-10) Agglutinating Sera are produced in rabbits and preserved with 0.5% phenol. Each dropper bottle contains 2 mL liquid and is supplied ready to use.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
Specifications
Specifications
| Content And Storage | 2°C to 8°C |
| Description | Shigella dysenteriae Polyvalent (1-10) |
| CE Marker | Yes |
| Detectable Analytes | Shigella |
| Format | Vial |
| Packaging Type | 2 mL/Vial |
| Quantity | 2 mL |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.