Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Shigella flexneri Polyvalent (1 to 6, X and Y) Agglutinating Sera

Catalog No. R30163801 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R30163801 2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. R30163801 Supplier Thermo Scientific™ Supplier No. R30163801
Add to Cart
Add to Cart
This item is not returnable. View return policy

Identify Shigella cultures presumptively with slide agglutination technique using Shigella flexneri Polyvalent (1 to 6, X and Y) Agglutinating Sera.

Presumptively identify Shigella cultures for epidemiological and diagnostic purposes using Thermo Scientific™ Remel™ Shigella flexneri Polyvalent (1 to 6, X and Y) Agglutinating Sera. These sera are suitable for use in slide agglutination tests. Serological tests are based on the fact that antibodies produced in the serum in response to exposure to bacterial antigens will visibly agglutinate with bacteria carrying homologous antigens. Antisera provide serological identification only; full identification of an organism must be made in conjunction with biochemical testing.

Shigella flexneri can be differentiated on the basis of their reactions with specific antisera. The Remel range of polyvalent antisera cover S. flexneri types 1 to 6, X and Y. As Shigella can be confused with some non-motile serotypes of E. coli (Alkalescens‑Dispar group) an additional serum (R30164301) is available for differentiation.

  • Ready to use: Supplied as ready to use agglutinating serum.
  • Easy to interpret: Visible agglutination with S. flexneri types 1 to 6, X and Y. Control solution shows no visible agglutination.

Shigella flexneri Polyvalent (1 to 6, X and Y) Agglutinating Sera are produced in rabbits and preserved with 0.5% phenol. Each dropper bottle contains 2 mL liquid and is supplied ready to use.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2°C to 8°C
Description Shigella flexneri Polyvalent (1 to 6, X and Y)
CE Marker Yes
Detectable Analytes Shigella
Format Vial
Packaging Type 2 mL/Vial
Quantity 2 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.