Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Vibrio cholerae Polyvalent Agglutinating Sera

Catalog No. r30165001
Change view
Click to view available options
Quantity:
2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R30165001 2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R30165001 Supplier Thermo Scientific™ Supplier No. R30165001
Only null left
Add to Cart
Add to Cart

Use Vibrio cholerae Polyvalent Agglutinating Serum for serological identification of V. cholerae O1 for epidemiological and diagnostic purposes.

Serologically identify Vibrio cholerae O1 by slide agglutination technique for epidemiological and diagnostic purposes using Thermo Scientific™ Remel™ Vibrio cholerae Polyvalent Agglutinating Serum. Serological tests are based on the fact that antibodies produced in the serum in response to exposure to bacterial antigens will visibly agglutinate with bacteria carrying homologous antigens.

Vibrio cholerae serogroup O1 is associated with most epidemic outbreaks of cholera. The O-serogrouping is based on antibodies that recognize structural differences in the heat stable O-antigen (lipopolysaccharide) of the bacteria. V. cholerae O1 and V. cholerae non-O1 isolates can be distinguished on the basis of simple agglutination tests.

  • Ready to use - Prepared reagent
  • Easy to store - Store at 2-8° C
  • Easy to interpret -
    • Test suspension - Agglutination should be strong and clearly visible within one minute.
    • Control suspension - No visible agglutination.

Vibrio cholerae Polyvalent Agglutinating Sera provides 2mL of rabbit serum containing antibodies to Vibrio cholerae, preserved with 0.5% phenol and supplied ready to use in a dropper bottle. Each dropper bottle contains sufficient sera for 40 to 50 tests.

This polyvalent serum is intended for use in slide agglutination screening tests.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

 

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2°C to 8°C
Description Vibrio cholerae Polyvalent
CE Marker Yes
Detectable Analytes V. cholerae
Format Vial
Packaging Type 2 mL/Vial
Quantity 2 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.