Learn More
Description
Thermo Scientific™ Remel Alkaline Peptone Water (pH 8.4) is used for cultivation of Vibrio.
Specifications
Specifications
| Description | Alkaline Peptone Water; pH 8.4 |
| Format | 15mm x 103mm Tube |
| Product Type | Buffer Solution |
| Quantity | 20 x 5 mL |
| Yield | 20/Pk. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.