Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Remel™ Alsevers Sheep Blood

Catalog No. p-4521198
Change view
Click to view available options
Quantity:
50 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R54048 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. R54048 Supplier Thermo Scientific™ Supplier No. R54048
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Be confident that your animal blood meets the highest standards possible by using Remel Alsevers Sheep Blood, sourced from our own technologically-advanced sheep farm.

Be confident that your animal blood meets the highest standards possible by using Remel Alsevers Sheep Blood, sourced from our own technologically-advanced sheep farm.

TRUSTED_SUSTAINABILITY

Specifications

Species Sheep
Form Liquid
Additive Alsevers
Product Type Whole Blood
Quantity 50 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.