Learn More
Thermo Scientific™ Anaerobic LKV Blood Agar
Catalog No. R01045 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
10/Pk.
100/Pk.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R01045 100/Pk.
R01044 10/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. R01045 Supplier Thermo Scientific™ Supplier No. R01045
Add to Cart
Add to Cart
This item is not returnable. View return policy
Isolate and cultivate anaerobic organisms, including fastidious strains with this selective, ready-to-use medium.
Isolate and cultivate anaerobic organisms, including fastidious strains with Thermo Scientific™ Remel™ Anaerobic Blood Agar ready prepared plates. Anaerobic Blood Agar serves as a nutritious medium supporting good growth and typical pigmentation of fastidious and slow growing anaerobes, as well as other anaerobes of clinical importance. Selective agents like kanamycin, vancomycin, neomycin, and paromomycin make the medium selective for fastidious, anaerobic bacteria from mixed populations by inhibiting facultative and aerobic gram-negative organisms.
  • Kanamycin and vancomycin inhibit gram-positive microorganisms and facultative anaerobic bacteria, and select for gram-negative bacilli
TRUSTED_SUSTAINABILITY
Specifications
Description Anaerobic LKV Blood Agar
Format 12mm x 85mm Monoplate
Product Type Agar Plate
Quantity 100/Pk.