Learn More
Thermo Scientific™ Anaerobic (CDC) Neomycin Blood Agar
Catalog No. R01047
Change view
Click to view available options
Format:
10mm x 100mm Spacesaver Plate
Catalog No. Format
R01047 10mm x 100mm Spacesaver Plate
1 options
Catalog No. R01047 Supplier Thermo Scientific™ Supplier No. R01047
Support the growth and typical pigmentation of fastidious, slow-growing and other clinically-important anaerobes with with this selective medium.
Support the growth and typical pigmentation of fastidious, slow-growing and other clinically-important anaerobes with Thermo Scientific™ Remel™ Anaerobic (CDC) Neomycin Blood Agar [The Centers for Disease Control and Prevention (CDC) formulation]1
Adding neomycin facilitates the isolation of anaerobic organisms, such as Bacteriodes and Clostridium spp. and inhibits enteric gram-negative bacilli.
  • Neomycin inhibits gram-negative Enterobacteriaceae
TRUSTED_SUSTAINABILITY
Specifications
Description Anaerobic Neomycin Blood Agar
Format 10mm x 100mm Spacesaver Plate
Product Type Agar Plate
Quantity 15/Pk.