Learn More
Description
Detect indole production and nitrate reduction in anaerobic bacteria using Thermo Scientific™ Remel™ Anaerobic Nitrate Reagent A. In 1977, Wideman et al. utilized a Nitrate Disk to determine nitrate reductase production and found 89% test agreement with the more conventional Indole Nitrate Medium1.
Specifications
Specifications
| Content And Storage | 2°C to 8°C |
| Description | Anaerobic Nitrate Reagent A |
| Detectable Analytes | Nitrite, Free Nitrogen Gas |
| Format | Bottle |
| Quantity | 25 mL |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.