Learn More
Thermo Scientific™ Remel™ Anaerobic Nitrate Reagent B
Catalog No. R21211 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
25 mL
Catalog No. Quantity
R21211 25 mL
1 options
Catalog No. R21211 Supplier Thermo Scientific™ Supplier No. R21211
Thermo Scientific™ Remel Anaerobic Nitrate Reagent B is for determination of nitrate reduction in anaerobic bacteria.
Thermo Scientific™ Remel Anaerobic Nitrate Reagent B is for determination of nitrate reduction in anaerobic bacteria.
Ordering Alerts:
Used in conjunction with Anaerobic Nitrate Reagent A, Part No. R21201
TRUSTED_SUSTAINABILITY
Specifications
Description Anaerobic Nitrate Reagent B
Detectable Analytes Nitrite, Free Nitrogen Gas
Format Bottle
Quantity 25 mL