Learn More
Thermo Scientific™ Anaerobic PEA Blood Agar (Anaerobic Phenylethyl Alcohol Agar) with 5% Sheep Blood
Shop All Thermo Scientific Products

Description
Isolate and cultivate anaerobic organisms, including fastidious strains with this selective, ready-to-use medium.
Isolate and cultivate anaerobic organisms, including fastidious strains with Thermo Scientific™ Remel™ Anaerobic Blood Agar ready prepared plates. Anaerobic Blood Agar serves as a nutritious medium supporting good growth and typical pigmentation of fastidious and slow growing anaerobes, as well as other anaerobes of clinical importance. Selective agents like kanamycin, vancomycin, neomycin and paramomycin make the medium selective for fastidious, anaerobic bacteria from mixed populations by inhibiting facultative and aerobic gram-negative organisms.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Description | Anaerobic PEA Blood Agar |
| Format | 12mm x 85mm Monoplate |
| Product Type | Agar Plate |
| Quantity | 100/Pk. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.