Description
Cultivate and isolate fastidious anaerobic bacteria with Thermo Scientific™ Remel™ Anaerobic Reducible Blood Agar. Adding reducing agents lowers the oxidation-reduction potential of the medium and improves the recovery of anaerobic organisms1
Anaerobic Reducible Blood Agar is a solid media recommended for qualitative primary isolation and cultivation procedures used for fastidious anaerobic bacteria.
Anaerobic Reducible Blood Agar is a nutritious medium containing peptone, hemin, vitamin K and sheep blood, which supply anaerobic organisms with required growth factors.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Description | Anaerobic Reducible Blood Agar |
| Format | 12mm x 85mm Monoplate |
| Product Type | Agar Plate |
| Quantity | 10/Pk. |