Learn More
Thermo Scientific™ Anaerobic Reducible Blood Agar with colistin and nalidixic acid (CNA)
Shop All Thermo Scientific Products
Description
Cultivate and isolate fastidious anaerobic bacteria with Thermo Scientific™ Remel™ Anaerobic Reducible Blood Agar.
The addition of cysteine, palladium chloride, and dithiothreitol makes the medium reducible. Reducible medium improves the recovery of anaerobes by lowering the oxidation – reduction potential of the medium. Kanamycin and vancomycin are added as selective agents for recovery of Gram-negative anaerobes, especially Bacteriodes spp
- Colistin and nalidixic acid inhibit gram-negative bacilli
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Description | Anaerobic Reducible; CNA Agar Base |
| Format | 12mm x 85mm Monoplate |
| Product Type | Agar Plate |
| Quantity | 10/Pk. |