Description
Cultivate and isolate fastidious anaerobic bacteria with Thermo Scientific™ Remel™ Anaerobic Reducible LKV Blood Agar. Adding reducing agents lowers the oxidation-reduction potential of the medium and improves the recovery of anaerobic organisms1. Kanamycin and vancomycin are added as selective agents for recovery of gram-negative anaerobes, especially Bacteriodes spp2.
- Kanamycin and vancomycin inhibit gram-positive microorganisms and facultative anaerobes
Specifications
Specifications
| Description | Anaerobic Reducible LKV Blood Agar |
| Format | 12mm x 85mm Monoplate |
| Product Type | Agar Plate |
| Quantity | 10/Pk. |